Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BI-78D3 | 883065-90-5 | MFCD01313575 | >98.0% | 379.37 | C13H9N5O5S2 | 1 ML
SDP

Catalog No. 5000245097
Encompass_Preferred

BI-78D3 (HY-10366) is a small-molecule substrate-competitive inhibitor of c-Jun N-terminal kinase (JNK) used in laboratory research. It inhibits JNK kinase activity with a reported IC50 of approximately 280 nM, and has molecular formula C13H9N5O5S2 and molecular weight 379.37 g/mol. The compound is supplied for research use only and is available in solid or solution formats.

  • Substrate-competitive JNK inhibitor.
  • Reported IC50 ≈ 280 nM against JNK.
  • Molecular formula C13H9N5O5S2.
  • Molecular weight 379.37 g/mol.
  • Supplied for research use only (not for human or veterinary use).
  • Available in solid or solution formats from distributors.

Catalog No. 50-002-45097 Supplier Medchemexpress LLC Supplier No. HY1036610MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BI-78D3 (HY-10366) is a small-molecule substrate-competitive inhibitor of c-Jun N-terminal kinase (JNK) used in laboratory research. It inhibits JNK kinase activity with a reported IC50 of approximately 280 nM, and has molecular formula C13H9N5O5S2 and molecular weight 379.37 g/mol. The compound is supplied for research use only and is available in solid or solution formats.

  • Substrate-competitive JNK inhibitor.
  • Reported IC50 ≈ 280 nM against JNK.
  • Molecular formula C13H9N5O5S2.
  • Molecular weight 379.37 g/mol.
  • Supplied for research use only (not for human or veterinary use).
  • Available in solid or solution formats from distributors.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.