Learn More
Medchemexpress LLC Bi-78d3 | 883065-90-5 | MFCD12912396 | 99.9% | 379.37 g/mol | C13H9N5O5S2 | 10 MG
SDP
Catalog No. 5000246834
Encompass_Preferred
BI-78D3 is a substrate-competitive inhibitor of c-Jun N-terminal kinase (JNK) used as a research tool to probe JNK-mediated signaling. It inhibits JNK kinase activity with an IC50 of approximately 280 nM and is supplied in solid and solution formats for in vitro and cell-based studies. For research use only.
  • Substrate-competitive inhibitor of JNK.
  • Reported IC50 of 280 nM against JNK.
  • High purity (99.85%).
  • Available as solid (10 mg) and as 10 mM solution in DMSO.
  • Powder storage: -20°C for long-term stability.
Catalog No. 50-002-46834 Supplier Medchemexpress LLC Supplier No. HY1036610MG
Add to Cart
Add to Cart
BI-78D3 is a substrate-competitive inhibitor of c-Jun N-terminal kinase (JNK) used as a research tool to probe JNK-mediated signaling. It inhibits JNK kinase activity with an IC50 of approximately 280 nM and is supplied in solid and solution formats for in vitro and cell-based studies. For research use only.
  • Substrate-competitive inhibitor of JNK.
  • Reported IC50 of 280 nM against JNK.
  • High purity (99.85%).
  • Available as solid (10 mg) and as 10 mM solution in DMSO.
  • Powder storage: -20°C for long-term stability.