Learn More
Apexbio Technology LLC BI-847325 1207293-36-4 100mg
SDP
Catalog No. 5000087796
Encompass_Preferred
BI-847325 (CAS 1207293-36-4) is a dual inhibitor targeting MEK1/2 and Aurora kinases with reported IC50 values of 25 nM and 4 nM against MEK1 and MEK2 and 3 nM 25 nM and 15 nM against Aurora A B and C respectively By inhibiting these kinases BI-847325 disrupts the MAPK/ERK and cell cycle signaling pathways that regulate cell proliferation and survival In cellular assays it demonstrates nanomolar EC50 values for inhibition of pERK and pHH3 in KRAS- and BRAF-mutant cancer cell lines and potently suppresses proliferation across a wide range of tumor cell models In xenograft studies it fully blocks tumor growth in BRAF- and KRAS-mutant cancer models This compound is utilized in research on kinase-targeted cancer therapies
Catalog No. 50-000-87796 Supplier Apexbio Technology LLC Supplier No. B6014100
BI-847325 (CAS 1207293-36-4) is a dual inhibitor targeting MEK1/2 and Aurora kinases with reported IC50 values of 25 nM and 4 nM against MEK1 and MEK2 and 3 nM 25 nM and 15 nM against Aurora A B and C respectively By inhibiting these kinases BI-847325 disrupts the MAPK/ERK and cell cycle signaling pathways that regulate cell proliferation and survival In cellular assays it demonstrates nanomolar EC50 values for inhibition of pERK and pHH3 in KRAS- and BRAF-mutant cancer cell lines and potently suppresses proliferation across a wide range of tumor cell models In xenograft studies it fully blocks tumor growth in BRAF- and KRAS-mutant cancer models This compound is utilized in research on kinase-targeted cancer therapies