Learn More
Medchemexpress LLC Bi-847325 | 1207293-36-4 | 98.2% | 10 MG
SDP
Catalog No. 5000295149
Encompass_Preferred
BI-847325 is an orally bioavailable, ATP-competitive dual inhibitor of MEK and Aurora kinases used in preclinical research to probe MAPK signaling and cell-cycle regulation. The compound has documented biochemical potency and is supplied as a solid research reagent for in vitro and in vivo studies.
  • Dual MEK and Aurora kinase inhibition for pathway and anticancer research.
  • Demonstrated in vitro and in vivo activity supporting translational studies.
  • High purity suitable for research applications.
  • Solid, light yellow to yellow appearance for easy handling.
  • Stable under recommended storage conditions to preserve compound integrity.
  • Provided as a small-scale research quantity for experimental use.
Catalog No. 50-002-95149 Supplier Medchemexpress LLC Supplier No. HY1895510MG
BI-847325 is an orally bioavailable, ATP-competitive dual inhibitor of MEK and Aurora kinases used in preclinical research to probe MAPK signaling and cell-cycle regulation. The compound has documented biochemical potency and is supplied as a solid research reagent for in vitro and in vivo studies.
  • Dual MEK and Aurora kinase inhibition for pathway and anticancer research.
  • Demonstrated in vitro and in vivo activity supporting translational studies.
  • High purity suitable for research applications.
  • Solid, light yellow to yellow appearance for easy handling.
  • Stable under recommended storage conditions to preserve compound integrity.
  • Provided as a small-scale research quantity for experimental use.