Learn More
Selleck Chemical LLC BI-847325
SDP
Catalog No. 5000001636
Encompass_Preferred
BI-847325 is an orally bioavailable and selective dual MEK/Aurora kinase inhibitor with IC50 of 3 nM 25 nM 15 nM 25 nM and 4 nM for Xenopus laevis Aurora B human Aurora A and Aurora C as well as human MEK1 and MEK2 respectively Phase 1
Catalog No. 50-000-01636 Supplier Selleck Chemical LLC Supplier No. S784325MG
BI-847325 is an orally bioavailable and selective dual MEK/Aurora kinase inhibitor with IC50 of 3 nM 25 nM 15 nM 25 nM and 4 nM for Xenopus laevis Aurora B human Aurora A and Aurora C as well as human MEK1 and MEK2 respectively Phase 1