Learn More
Medchemexpress LLC HY-18955 5mg Medchemexpress, BI-847325 CAS:1207293-36-4 Purity:>98%
SDP
Catalog No. 502082098
Encompass_Preferred
Medchemexpress, HY-18955 5mg BI-847325 CAS:1207293-36-4 BI-847325 is an ATP competitive dual inhibitor of MEK and aurora kinases (AK) with IC50 values of 4 and 15 nM for human MEK2 and AK-C, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2098 Supplier Medchemexpress LLC Supplier No. HY189555MG
Medchemexpress, HY-18955 5mg BI-847325 CAS:1207293-36-4 BI-847325 is an ATP competitive dual inhibitor of MEK and aurora kinases (AK) with IC50 values of 4 and 15 nM for human MEK2 and AK-C, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.