Learn More
TARGETMOL CHEMICALS INC BI-847325 5MG
SDP
Catalog No. 502880493
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. BI-847325 is a selective dual inhibitor of MEK and aurora kinases (AK) with IC50 values of 4 and 15 nM for human MEK2 and AK-C, respectively. Purity 97.13%
Catalog No. 50-288-0493 Supplier TARGETMOL CHEMICALS INC Supplier No. T67855MG
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. BI-847325 is a selective dual inhibitor of MEK and aurora kinases (AK) with IC50 values of 4 and 15 nM for human MEK2 and AK-C, respectively. Purity 97.13%