Learn More
TARGETMOL CHEMICALS INC BI-882370 10MG
SDP
Catalog No. 502877189
Encompass_Preferred
Also available in 1 mg, 2 mg, 5 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. BI-882370 is a specific RAF kinase inhibitor. BI-882370 inhibits the oncogenic BRAFV600E-mutant, the WT BRAF and CRAF kinases (IC50s 0.4, 0.8 and 0.6 nM). BI-882370 also inhibits SRC family kinases. Purity 99.07%
Catalog No. 50-287-7189 Supplier TARGETMOL CHEMICALS INC Supplier No. TQ004810MG
Also available in 1 mg, 2 mg, 5 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. BI-882370 is a specific RAF kinase inhibitor. BI-882370 inhibits the oncogenic BRAFV600E-mutant, the WT BRAF and CRAF kinases (IC50s 0.4, 0.8 and 0.6 nM). BI-882370 also inhibits SRC family kinases. Purity 99.07%