Learn More
Medchemexpress LLC BI-9564 | 1883429-22-8 | MFCD09930810 | 98.7% | 353.41 | 5 MG
SDP
Catalog No. 5000364482
Encompass_Preferred
BI-9564 is a potent, selective, and cell-permeable BRD9/BRD7 bromodomains inhibitor.
  • Potent, selective, and cell-permeable BRD9/BRD7 bromodomains inhibitor.
  • IC₅₀ of 75 nM for BRD9 and 3.4 μM for BRD7.
  • Kds of 14 nM for BRD9 and 239 nM for BRD7.
  • Highly selective, with an IC₅₀ > 100 μM for BET family.
  • Shows no activity against 324 kinases (<5 μM).
  • Antiproliferative effect on human acute myeloid eosinophilic leukemia cell line EOL-1 (EC₅₀ 800 nM).
  • More than 10-fold selective for BRD9 over the highly homologous bromodomain BRD7.
  • Attractive ADME/PK profiles for in vivo proof-of-concept studies.
  • Results in additional survival benefit in a xenograft model of human AML.
Catalog No. 50-003-64482 Supplier Medchemexpress LLC Supplier No. 5MGHY100352
BI-9564 is a potent, selective, and cell-permeable BRD9/BRD7 bromodomains inhibitor.
  • Potent, selective, and cell-permeable BRD9/BRD7 bromodomains inhibitor.
  • IC₅₀ of 75 nM for BRD9 and 3.4 μM for BRD7.
  • Kds of 14 nM for BRD9 and 239 nM for BRD7.
  • Highly selective, with an IC₅₀ > 100 μM for BET family.
  • Shows no activity against 324 kinases (<5 μM).
  • Antiproliferative effect on human acute myeloid eosinophilic leukemia cell line EOL-1 (EC₅₀ 800 nM).
  • More than 10-fold selective for BRD9 over the highly homologous bromodomain BRD7.
  • Attractive ADME/PK profiles for in vivo proof-of-concept studies.
  • Results in additional survival benefit in a xenograft model of human AML.