Learn More
Selleck Chemical LLC BI-9564
SDP
Catalog No. 5000001392
Encompass_Preferred
BI-9564 is a selective inhibitor of BRD9 and BRD7 bromodomains with the IC50 of 75 nM and 3 4 M respectively
Catalog No. 50-000-01392 Supplier Selleck Chemical LLC Supplier No. S811325MG
BI-9564 is a selective inhibitor of BRD9 and BRD7 bromodomains with the IC50 of 75 nM and 3 4 M respectively