Learn More
Medchemexpress LLC BI-9627 | 1204329-34-9 | 99.7% | 356.34 Da | C16H19F3N4O2 | 10 MG
SDP
Catalog No. 5000295368
Encompass_Preferred
BI-9627 is a potent small-molecule inhibitor of the sodium-hydrogen exchanger isoform 1 (NHE1), used as a research chemical probe to study NHE1-mediated physiology and pharmacology. The compound is supplied as a solid, has a reported molecular formula of C16H19F3N4O2 and a molecular weight of about 356.34 Da, with high reported purity suitable for biochemical and cellular assays.
  • Potent NHE1 inhibitor with low-nanomolar activity in cell-based assays
  • High chemical purity suitable for biochemical and cellular studies
  • Supplied as a solid for ease of handling and storage
  • Molecularly characterized with CAS and formula for unambiguous identification
  • Useful as a chemical probe for studying sodium-hydrogen exchange pathways
Catalog No. 50-002-95368 Supplier Medchemexpress LLC Supplier No. HY1807110MG
BI-9627 is a potent small-molecule inhibitor of the sodium-hydrogen exchanger isoform 1 (NHE1), used as a research chemical probe to study NHE1-mediated physiology and pharmacology. The compound is supplied as a solid, has a reported molecular formula of C16H19F3N4O2 and a molecular weight of about 356.34 Da, with high reported purity suitable for biochemical and cellular assays.
  • Potent NHE1 inhibitor with low-nanomolar activity in cell-based assays
  • High chemical purity suitable for biochemical and cellular studies
  • Supplied as a solid for ease of handling and storage
  • Molecularly characterized with CAS and formula for unambiguous identification
  • Useful as a chemical probe for studying sodium-hydrogen exchange pathways