Learn More
Medchemexpress LLC BI-9627 (hydrochloride) | 1422252-46-7 | 98.0% | 392.80 | 1 ML
SDP
Catalog No. 5000387406
Encompass_Preferred
BI-9627 hydrochloride is a potent and selective sodium-hydrogen exchanger isoform 1 (NHE1) inhibitor, demonstrating over 30-fold selectivity against NHE2. It has been shown to decrease autophagy in HTR-8/SVneo cells, significantly reduce the pHi of human sperm, and prolong Ca2+ recovery time in KO hiPSC-CMs. This compound exhibits low drug-drug interaction potential, excellent pharmacokinetics in rat and dog models, and potent activity in isolated heart models of ischemia-reperfusion injury.
  • Potent sodium-hydrogen exchanger isoform 1 (NHE1) inhibitor
  • Over 30-fold selectivity against NHE2
  • Decreases autophagy in HTR-8/SVneo cells
  • Significantly reduces pHi of human sperm
  • Prolongs Ca2+ recovery time in KO hiPSC-CMs
  • Low drug-drug interaction potential
  • Excellent pharmacokinetics in rat and dog models
  • Potent activity in isolated heart models of ischemia-reperfusion injury
Catalog No. 50-003-87406 Supplier Medchemexpress LLC Supplier No. HY1807110MM/1ML
BI-9627 hydrochloride is a potent and selective sodium-hydrogen exchanger isoform 1 (NHE1) inhibitor, demonstrating over 30-fold selectivity against NHE2. It has been shown to decrease autophagy in HTR-8/SVneo cells, significantly reduce the pHi of human sperm, and prolong Ca2+ recovery time in KO hiPSC-CMs. This compound exhibits low drug-drug interaction potential, excellent pharmacokinetics in rat and dog models, and potent activity in isolated heart models of ischemia-reperfusion injury.
  • Potent sodium-hydrogen exchanger isoform 1 (NHE1) inhibitor
  • Over 30-fold selectivity against NHE2
  • Decreases autophagy in HTR-8/SVneo cells
  • Significantly reduces pHi of human sperm
  • Prolongs Ca2+ recovery time in KO hiPSC-CMs
  • Low drug-drug interaction potential
  • Excellent pharmacokinetics in rat and dog models
  • Potent activity in isolated heart models of ischemia-reperfusion injury