Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BI-9627 hydrochloride | 1422252-46-7 | 98.0% | 392.80 | 5 MG
SDP

Catalog No. 5000387453
Encompass_Preferred

BI-9627 hydrochloride | 1422252-46-7 | 98.0% | 392.80 | 5 MG

Catalog No. 50-003-87453 Supplier Medchemexpress LLC Supplier No. HY18071A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

BI-9627 hydrochloride | 1422252-46-7 | 98.0% | 392.80 | 5 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.