Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BI-9627 hydrochloride | 1422252-46-7 | 98.0% | 392.80 g/mol | C16H20ClF3N4O2 | 10 MG
SDP

Catalog No. 5000297969
Encompass_Preferred

BI-9627 hydrochloride is the hydrochloride salt of a potent sodium-hydrogen exchanger 1 (NHE1) inhibitor supplied for research use. The compound is intended for biochemical and pharmacological studies of NHE1 activity and is accompanied by analytical documentation to support quality and identity.

  • Potent NHE1 inhibition at low nanomolar concentrations.
  • Hydrochloride salt form improves solubility for formulation and handling.
  • High purity suitable for research applications and analytical studies.
  • Characterized molecular weight 392.80 g/mol and formula C16H20ClF3N4O2.
  • Supplied with supporting documentation such as COA and analytical data.

Catalog No. 50-002-97969 Supplier Medchemexpress LLC Supplier No. HY18071A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

BI-9627 hydrochloride is the hydrochloride salt of a potent sodium-hydrogen exchanger 1 (NHE1) inhibitor supplied for research use. The compound is intended for biochemical and pharmacological studies of NHE1 activity and is accompanied by analytical documentation to support quality and identity.

  • Potent NHE1 inhibition at low nanomolar concentrations.
  • Hydrochloride salt form improves solubility for formulation and handling.
  • High purity suitable for research applications and analytical studies.
  • Characterized molecular weight 392.80 g/mol and formula C16H20ClF3N4O2.
  • Supplied with supporting documentation such as COA and analytical data.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.