Learn More
Medchemexpress LLC Bi-9627 (hydrochloride) | 1422252-46-7 | 98.01% | 392.80 | 1 ML
SDP
Catalog No. 5000387248
Encompass_Preferred
BI-9627 hydrochloride is a potent inhibitor of sodium-hydrogen exchanger isoform 1 (NHE1), exhibiting high selectivity over NHE2 and no measurable activity against NHE3. This compound shows low agent-agent interaction potential, excellent pharmacokinetics, and potent activity in isolated heart models of ischemia-reperfusion injury. It is intended for research use only.
  • Decreases autophagy in HTR-8/SVneo cells.
  • Significantly reduces the pHi of human sperm and partially reverses the effect of DMA.
  • Prolongs Ca2+ recovery time in KO hiPSC-CMs.
  • Shows low DDI potential.
  • Excellent pharmacokinetics in rat and dog.
  • Potent activity in isolated heart models of ischemia-reperfusion injury.
Catalog No. 50-003-87248 Supplier Medchemexpress LLC Supplier No. HY18071A10MM/1ML
BI-9627 hydrochloride is a potent inhibitor of sodium-hydrogen exchanger isoform 1 (NHE1), exhibiting high selectivity over NHE2 and no measurable activity against NHE3. This compound shows low agent-agent interaction potential, excellent pharmacokinetics, and potent activity in isolated heart models of ischemia-reperfusion injury. It is intended for research use only.
  • Decreases autophagy in HTR-8/SVneo cells.
  • Significantly reduces the pHi of human sperm and partially reverses the effect of DMA.
  • Prolongs Ca2+ recovery time in KO hiPSC-CMs.
  • Shows low DDI potential.
  • Excellent pharmacokinetics in rat and dog.
  • Potent activity in isolated heart models of ischemia-reperfusion injury.