Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BI-9787 | 3043939-18-7 | 99.5% | 489.58 | 100 MG
SDP

Catalog No. 5000394835
Encompass_Preferred

BI-9787 is a zwitterionic inhibitor for ketohexokinase, with an IC50 of 12 nM for hKHK-A and 12.8 nM for hKHK-C. It shows good metabolic stability in rat hepatocytes and good pharmacokinetic characteristics in rats. The compound was designed by Boehringer Ingelheim and is available through the opnMe.com open innovation portal, along with its negative control. It is intended for research use only and not for sale to patients.

  • Purity: 99.46%
  • CAS number: 3043939-18-7
  • Molecular weight: 489.58
  • Formula: C24H29F2N5O2S
  • Appearance: Solid
  • Color: White to off-white
  • IC50 and target: 12 nM (hKHK-A); 12.8 nM (hKHK-C)

Catalog No. 50-003-94835 Supplier Medchemexpress LLC Supplier No. HY161993100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BI-9787 is a zwitterionic inhibitor for ketohexokinase, with an IC50 of 12 nM for hKHK-A and 12.8 nM for hKHK-C. It shows good metabolic stability in rat hepatocytes and good pharmacokinetic characteristics in rats. The compound was designed by Boehringer Ingelheim and is available through the opnMe.com open innovation portal, along with its negative control. It is intended for research use only and not for sale to patients.

  • Purity: 99.46%
  • CAS number: 3043939-18-7
  • Molecular weight: 489.58
  • Formula: C24H29F2N5O2S
  • Appearance: Solid
  • Color: White to off-white
  • IC50 and target: 12 nM (hKHK-A); 12.8 nM (hKHK-C)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.