Learn More
Medchemexpress LLC BI-9787 | 3043939-18-7 | 99.5% | 489.58 | 5 MG
SDP
Catalog No. 5000395286
Encompass_Preferred
BI-9787 is a zwitterionic inhibitor for ketohexokinase (KHK), demonstrating IC50 values of 12 nM for hKHK-A and 12.8 nM for hKHK-C. It shows good metabolic stability in rat hepatocytes and favorable pharmacokinetic characteristics in rats. This product was designed by Boehringer Ingelheim and is available free of charge through their open innovation portal opnMe.com, alongside its negative control. It is intended for research use only and not for sale to patients.
  • Zwitterionic inhibitor for ketohexokinase (KHK)
  • IC50 of 12 nM for hKHK-A and 12.8 nM for hKHK-C
  • Exhibits good metabolic stability in rat hepatocyte
  • Shows good pharmacokinetic characteristics in rats
  • Intended for research use only
Catalog No. 50-003-95286 Supplier Medchemexpress LLC Supplier No. HY1619935MG
BI-9787 is a zwitterionic inhibitor for ketohexokinase (KHK), demonstrating IC50 values of 12 nM for hKHK-A and 12.8 nM for hKHK-C. It shows good metabolic stability in rat hepatocytes and favorable pharmacokinetic characteristics in rats. This product was designed by Boehringer Ingelheim and is available free of charge through their open innovation portal opnMe.com, alongside its negative control. It is intended for research use only and not for sale to patients.
  • Zwitterionic inhibitor for ketohexokinase (KHK)
  • IC50 of 12 nM for hKHK-A and 12.8 nM for hKHK-C
  • Exhibits good metabolic stability in rat hepatocyte
  • Shows good pharmacokinetic characteristics in rats
  • Intended for research use only