Learn More
Medchemexpress LLC BI-9787 is a zwitterionic inhibitor for ketohexokinase. | 3043939-18-7 | 99.5% | 489.58 | 5 MG
SDP
Catalog No. 5000395090
Encompass_Preferred
BI-9787 is a zwitterionic inhibitor for ketohexokinase (KHK), with an IC50 of 12 nM for hKHK-A and 12.8 nM for hKHK-C. It demonstrates good metabolic stability in rat hepatocytes and favorable pharmacokinetic characteristics in rats.
  • Exhibits good metabolic stability in rat hepatocyte.
  • Demonstrates good pharmacokinetic characteristics in rats.
Catalog No. 50-003-95090 Supplier Medchemexpress LLC Supplier No. HY16199350MG
BI-9787 is a zwitterionic inhibitor for ketohexokinase (KHK), with an IC50 of 12 nM for hKHK-A and 12.8 nM for hKHK-C. It demonstrates good metabolic stability in rat hepatocytes and favorable pharmacokinetic characteristics in rats.
  • Exhibits good metabolic stability in rat hepatocyte.
  • Demonstrates good pharmacokinetic characteristics in rats.