Learn More
Medchemexpress LLC BI 99179 | 1291779-76-4 | 99.38% | 391.46 | 5 MG
SDP
Catalog No. 5000364366
Encompass_Preferred
BI 99179 is a chemical probe that acts as a potent and selective type I fatty acid synthase (FAS) inhibitor with an IC50 of 79 nM. It is a tool compound suitable for in vivo validation of FAS as a target for lipid metabolism-related diseases and shows significant peripheral and central exposure upon oral administration in rats.
  • Potent in mouse hypothalamic N-42 cells with an IC50 of 0.6 μM.
  • No significant LDH release in cytotoxicity assays up to 30 μM.
  • Exhibits antiproliferative efficacy in human glioma GAMG cells.
  • Possesses a super pharmacokinetic profile in male Han/Wistar rats.
  • For research use only.
Catalog No. 50-003-64366 Supplier Medchemexpress LLC Supplier No. 5MGHY16100
BI 99179 is a chemical probe that acts as a potent and selective type I fatty acid synthase (FAS) inhibitor with an IC50 of 79 nM. It is a tool compound suitable for in vivo validation of FAS as a target for lipid metabolism-related diseases and shows significant peripheral and central exposure upon oral administration in rats.
  • Potent in mouse hypothalamic N-42 cells with an IC50 of 0.6 μM.
  • No significant LDH release in cytotoxicity assays up to 30 μM.
  • Exhibits antiproliferative efficacy in human glioma GAMG cells.
  • Possesses a super pharmacokinetic profile in male Han/Wistar rats.
  • For research use only.