Learn More
Alpco Ltd Bi-CAT (Epinephrine & Norepinephrine) ELISA, 2 plate
SDP
Supplier:  Alpco Ltd 17BCTHUE02.1
Encompass
The Bi-CAT ELISA (Epinephrine & Norepinephrine ELISA) is a two-plate enzyme Immunoassay for the quantitative determination of adrenaline (epinephrine) and noradrenaline (norepinephrine) in plasma and urine.
For Research Use Only. Not for Use in Diagnostic Procedures. Includes 2 plates.
Catalog No. 50-751-3487