Learn More
Medchemexpress LLC (±)-BI-D | 1416258-16-6 | 98.06% | 405.49 | 50 MG
SDP
Catalog No. 5000387784
Encompass_Preferred
(±)-BI-D is a potent allosteric integrase inhibitor (ALLINI). It operates by binding to integrase at the LEDGF/p75 binding site, effectively inhibiting HIV-Luc infection in cellular assays.
  • Potent allosteric integrase inhibitor.
  • Binds integrase at the LEDGF/p75 binding site.
  • Inhibits HIV-Luc infection in cells.
  • For research use only.
Catalog No. 50-003-87784 Supplier Medchemexpress LLC Supplier No. HY1860150MG
(±)-BI-D is a potent allosteric integrase inhibitor (ALLINI). It operates by binding to integrase at the LEDGF/p75 binding site, effectively inhibiting HIV-Luc infection in cellular assays.
  • Potent allosteric integrase inhibitor.
  • Binds integrase at the LEDGF/p75 binding site.
  • Inhibits HIV-Luc infection in cells.
  • For research use only.