Learn More
Medchemexpress LLC (±)-BI-D | 1416258-16-6 | 98.1% | 10 MG
SDP
Catalog No. 5000295466
Encompass_Preferred
(±)-BI-D is an allosteric HIV integrase inhibitor used in research to study integrase function and antiviral activity. It binds the LEDGF/p75 binding site on integrase and has demonstrated inhibition of HIV-Luc infection in cellular assays.
  • Potent ALLINI mechanism of action.
  • Binds LEDGF/p75 integrase site to disrupt viral integration.
  • Demonstrated cellular activity (reported IC50s: 0.16 μM and 2.9 μM).
  • Available as solid and as a 10 mM solution in DMSO for assay setup flexibility.
  • High purity and defined molecular weight and formula for reproducible studies.
  • Intended for biochemical and cellular research use only; not for human use.
Catalog No. 50-002-95466 Supplier Medchemexpress LLC Supplier No. HY1860110MG
(±)-BI-D is an allosteric HIV integrase inhibitor used in research to study integrase function and antiviral activity. It binds the LEDGF/p75 binding site on integrase and has demonstrated inhibition of HIV-Luc infection in cellular assays.
  • Potent ALLINI mechanism of action.
  • Binds LEDGF/p75 integrase site to disrupt viral integration.
  • Demonstrated cellular activity (reported IC50s: 0.16 μM and 2.9 μM).
  • Available as solid and as a 10 mM solution in DMSO for assay setup flexibility.
  • High purity and defined molecular weight and formula for reproducible studies.
  • Intended for biochemical and cellular research use only; not for human use.