Learn More
Medchemexpress LLC (±)-BI-D | 1416258-16-6 | 98.1% | 405.49 | 10 MM * 1 ML
SDP
Catalog No. 5000388005
Encompass_Preferred
(±)-BI-D is a potent allosteric integrase inhibitor (ALLINI). It operates by binding to integrase at the LEDGF/p75 binding site. This compound has demonstrated inhibitory effects on HIV-Luc infection in cells, with an IC50 of 0.16 μM in Psip1 knockout E9 mouse embryonic fibroblasts and 2.9 μM in wild-type E9 mouse embryonic fibroblasts.
  • Potent ALLINI (allosteric integrase inhibitor)
  • Binds integrase at the LEDGF/p75 binding site
  • Inhibits HIV-Luc infection in cells
    • IC50: 0.16 μM in Psip1 knockout E9 mouse embryonic fibroblasts
    • IC50: 2.9 μM in wild-type E9 mouse embryonic fibroblasts
  • For research use only (not for sale to patients)
  • Available as a solid
  • Light yellow to yellow color
Catalog No. 50-003-88005 Supplier Medchemexpress LLC Supplier No. HY1860110MM/1ML
Add to Cart
Add to Cart
(±)-BI-D is a potent allosteric integrase inhibitor (ALLINI). It operates by binding to integrase at the LEDGF/p75 binding site. This compound has demonstrated inhibitory effects on HIV-Luc infection in cells, with an IC50 of 0.16 μM in Psip1 knockout E9 mouse embryonic fibroblasts and 2.9 μM in wild-type E9 mouse embryonic fibroblasts.
  • Potent ALLINI (allosteric integrase inhibitor)
  • Binds integrase at the LEDGF/p75 binding site
  • Inhibits HIV-Luc infection in cells
    • IC50: 0.16 μM in Psip1 knockout E9 mouse embryonic fibroblasts
    • IC50: 2.9 μM in wild-type E9 mouse embryonic fibroblasts
  • For research use only (not for sale to patients)
  • Available as a solid
  • Light yellow to yellow color