Learn More
Medchemexpress LLC (±)-BI-D | 1416258-16-6 | 98.1% | 405.49 | 100 MG
SDP
Catalog No. 5000387791
Encompass_Preferred
(±)-BI-D is a potent ALLINI (allosteric integrase inhibitor) that binds integrase at the LEDGF/p75 binding site. It inhibits HIV-Luc infection in cells, with an IC50 of 0.16 μM in Psip1 knockout E9 mouse embryonic fibroblasts and 2.9 μM in wild-type E9 mouse embryonic fibroblasts. The product is for research use only and not sold to patients.
  • Potent allosteric integrase inhibitor.
  • Binds integrase at the LEDGF/p75 binding site.
  • Inhibits HIV-Luc infection in cells.
Catalog No. 50-003-87791 Supplier Medchemexpress LLC Supplier No. HY18601100MG
(±)-BI-D is a potent ALLINI (allosteric integrase inhibitor) that binds integrase at the LEDGF/p75 binding site. It inhibits HIV-Luc infection in cells, with an IC50 of 0.16 μM in Psip1 knockout E9 mouse embryonic fibroblasts and 2.9 μM in wild-type E9 mouse embryonic fibroblasts. The product is for research use only and not sold to patients.
  • Potent allosteric integrase inhibitor.
  • Binds integrase at the LEDGF/p75 binding site.
  • Inhibits HIV-Luc infection in cells.