Learn More
Medchemexpress LLC HY-10510 10mg Medchemexpress, BI-D1870 CAS:501437-28-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1051010MG
Encompass_Preferred
Medchemexpress, HY-10510 10mg BI-D1870 CAS:501437-28-1 BI-D1870 is an ATP-competitive, cell permeable inhibitor of RSK isoforms, with IC50s of 31 nM/24 nM/18 nM/15 nM for RSK1/RSK2/RSK3/RSK4, respectively[2][3][4][5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8764