Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bi-D1870 | 501437-28-1 | MFCD11223662 | 99.4% | 391.42 g·mol⁻¹ | C19H23F2N5O2 | 1 ML
SDP

Catalog No. 5000243648
Encompass_Preferred

BI-D1870 is a cell-permeable, ATP-competitive inhibitor of p90 ribosomal S6 kinase (RSK) isoforms 1-4 that is used in biochemical and cell-based studies to probe RSK-dependent signaling. It preferentially targets the N-terminal kinase domain and has been reported to be brain penetrant.

  • ATP-competitive inhibitor of RSK isoforms (RSK1-4)
  • Potent IC50s: RSK1 31 nM; RSK2 24 nM; RSK3 18 nM; RSK4 15 nM
  • Cell permeable and reported to be brain penetrant
  • Acts at the N-terminal kinase domain of RSK
  • Available as solid or as a 10 mM solution in DMSO (1 mL)
  • Typical purity approximately 99.4%
  • Suitable for biochemical and cell-based studies of RSK signaling

Catalog No. 50-002-43648 Supplier Medchemexpress LLC Supplier No. HY1051010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BI-D1870 is a cell-permeable, ATP-competitive inhibitor of p90 ribosomal S6 kinase (RSK) isoforms 1-4 that is used in biochemical and cell-based studies to probe RSK-dependent signaling. It preferentially targets the N-terminal kinase domain and has been reported to be brain penetrant.

  • ATP-competitive inhibitor of RSK isoforms (RSK1-4)
  • Potent IC50s: RSK1 31 nM; RSK2 24 nM; RSK3 18 nM; RSK4 15 nM
  • Cell permeable and reported to be brain penetrant
  • Acts at the N-terminal kinase domain of RSK
  • Available as solid or as a 10 mM solution in DMSO (1 mL)
  • Typical purity approximately 99.4%
  • Suitable for biochemical and cell-based studies of RSK signaling

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.