Learn More
Enzo Life Sciences BI-D1870 (5mg). CAS: 501437-28-1
SDP
Supplier:  Enzo Life Sciences BMLEI4070005
Encompass_Preferred
Cell permeable, ATP-competitive inhibitor of the four isoforms of ribosomal S6 kinase (p90 RSK) (IC50's at 100µM ATP: RSK1 = 31nM, RSK2 = 24nM, RSK3 = 18nM, RSK4 = 15nM). Does not significantly inhibit 10 other AGC kinases, and over 40 other protein kinases at 100X concentrations. Acts on the N-terminal AGC kinase domain. Completely prevents PMA-induced TSC2 phosphorylation. Alternative name: 2-[(3,5-Difluoro-4-hydroxyphenyl)amino]-7,8-dihydro-5,7-dimethyl-8-(3-methylbutyl)-6(5H)-pteridone. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (>25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0784