Learn More
TARGETMOL CHEMICALS INC BI-D1870 5MG
SDP
Catalog No. 502576582
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BI-D1870 is a cell-permeable ATP-competitive inhibitor of ribosomal S6 kinases (RSKs; IC50s 31/24/18/15 nM for RSK1/2/3/4). purity: 99%
Catalog No. 50-257-6582 Supplier TARGETMOL CHEMICALS INC Supplier No. T61715MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BI-D1870 is a cell-permeable ATP-competitive inhibitor of ribosomal S6 kinases (RSKs; IC50s 31/24/18/15 nM for RSK1/2/3/4). purity: 99%