Learn More
Selleck Chemical LLC BI-D1870 5mg 501437-28-1
SDP
Catalog No. NC0426441
Encompass_Preferred
BI-D1870 is an ATP-competitive inhibitor of S6 ribosome for RSK1/2/3/4 with IC50 of 31 nM/24 nM/18 nM/15 nM in cell-free assays, respectively; 10- to 100-fold selectivity for RSK than MST2, GSK-3?, MARK3, CK1 and Aurora B. BI-D1870 exhibits anticancer attributes including the generation of reactive oxygen species (ROS) and increases in endoplasmic reticulum (ER) stress and autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0426441 Supplier Selleck Chemical LLC Supplier No. S28435MG
Add to Cart
Add to Cart
BI-D1870 is an ATP-competitive inhibitor of S6 ribosome for RSK1/2/3/4 with IC50 of 31 nM/24 nM/18 nM/15 nM in cell-free assays, respectively; 10- to 100-fold selectivity for RSK than MST2, GSK-3?, MARK3, CK1 and Aurora B. BI-D1870 exhibits anticancer attributes including the generation of reactive oxygen species (ROS) and increases in endoplasmic reticulum (ER) stress and autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.