Learn More
STEMCELL Technologies BI-D1870, Size: 10 mg
SDP
Supplier:  STEMCELL Technologies 72714
Encompass
BI-D1870 inhibits the 90 kDa ribosomal S6 kinases (RSKs), which are serine/threonine kinases involved in diverse cellular processes including growth, survival, and motility (Romeo et al.).MAINTENANCE AND SELF-RENEWAL· Reduces neural stem cell proliferation and self-renewal in vitro (Karelina et al.). CANCER RESEARCH· Inhibits growth of breast cancer cell lines (Stratford et al., Dhillon et al.).
Catalog No. 50-197-5967