Learn More
Thomson Instrument Company Bidirectional Transfer Cap, 2' Tubing w/ Luer Lock, Sterile
SDP
Supplier:  Thomson Instrument Company 9316188
Encompass_Preferred
Thomson’s Bidirectional Transfer Caps enable precise liquid handling in and out of bioreactors, cell bags, and Optimum Growth flasks—compatible with peristaltic pump setup. Designed for high-throughput facilities, these caps support flexible media exchange while maintaining a closed aseptic system.
• Bi-directional design with luer locks, tubing, or quick connects enables aseptic media transfer, inoculation, or sampling into or out of 1.6L, 2.8L, 5L, and 7L flasks
• Quick to assemble, reducing process downtime
• Sterile, closed-loop design with multiple connection options enables seamless integration into custom systems while maintaining aseptic integrity
• Flexible setup—compatible with common fluid transfer systems and pumps
• Made in the USA and individually packaged for cleanroom use
Catalog No. NC0930173