Learn More
Alkali Scientific Bi-directional Liquid Transfer
SDP
Catalog No. 503028573
Encompass_Preferred
Our 5L high efficiency shaker flasks are ideal for medium size scale up work They are inhigh efficiencyin in that a 5L flask has 230mm diameter footprint allowing for more of them to fit on a shaker platform The geometry of the flasks also allows for higher shaker speeds without damaging the cells Faster shaker speeds means better gas exchange higher growth rates and more yield per flask This product features pre-assembled tubing connectors and gas exchange filters allowing for the movement of liquid in and out of the Erlenmeyer flask with minimal risk of contamination Many customers have realized significant cost savings by switching to aseptic transfer systems 5L High Efficiency Erlenmeyer Flask 1 system per double wrapped sterile bag - 4 systems per case Flask made of USP grade VI virgin polycarbonate Untreated super smooth interior surface ideal for suspended cultures Attached bi-directional transfer cap Attached thermoplastic elastomer (TPE) external transfer tube No at
Catalog No. 50-302-8573 Supplier Alkali Scientific Supplier No. EFC720
Our 5L high efficiency shaker flasks are ideal for medium size scale up work They are inhigh efficiencyin in that a 5L flask has 230mm diameter footprint allowing for more of them to fit on a shaker platform The geometry of the flasks also allows for higher shaker speeds without damaging the cells Faster shaker speeds means better gas exchange higher growth rates and more yield per flask This product features pre-assembled tubing connectors and gas exchange filters allowing for the movement of liquid in and out of the Erlenmeyer flask with minimal risk of contamination Many customers have realized significant cost savings by switching to aseptic transfer systems 5L High Efficiency Erlenmeyer Flask 1 system per double wrapped sterile bag - 4 systems per case Flask made of USP grade VI virgin polycarbonate Untreated super smooth interior surface ideal for suspended cultures Attached bi-directional transfer cap Attached thermoplastic elastomer (TPE) external transfer tube No at