Learn More
Alkali Scientific Bi-directional Liquid Transfer Cap System for 3L High Efficiency Erlenmeyer Flasks, White, with TPE Tube (50cm 1/8" ID 1/4" OD), Vent Filter (0.22 μm Φ24mm), Sterile, 1/pk, 4/cs
SDP
Catalog No. 503028584
Encompass_Preferred
Our pre-installed on the High Efficiency Erlenmeyer Flask,which realizes the closed transfer of liquid and reduces the risk of external contami-nation during the process of capping and screwing. Closed system reduces the risks of contamination during liquid transfer. One-piece construction of caps and connectors reduces the risk of leakage and media residue. Caps of various diameters are available and aseptic welding of liquid inlet tubing under normal conditions is supported. High-quality materials and smooth inner wall of the tubing provide an excellent transfer performance. Sterilized by E-beam, SAL=10^-6. Endotoxin-free, and no ingredients of animal origin. Individually packaged in sterile bag.
Catalog No. 50-302-8584 Supplier Alkali Scientific Supplier No. EFC620
Our pre-installed on the High Efficiency Erlenmeyer Flask,which realizes the closed transfer of liquid and reduces the risk of external contami-nation during the process of capping and screwing. Closed system reduces the risks of contamination during liquid transfer. One-piece construction of caps and connectors reduces the risk of leakage and media residue. Caps of various diameters are available and aseptic welding of liquid inlet tubing under normal conditions is supported. High-quality materials and smooth inner wall of the tubing provide an excellent transfer performance. Sterilized by E-beam, SAL=10^-6. Endotoxin-free, and no ingredients of animal origin. Individually packaged in sterile bag.