Learn More
Chemglass Life Sciences Bi-Directional Liquid Transfer Cap, for NEST 2L High Efficiency Erlenmeyer Flasks
SDP
Catalog No. 502733453
Encompass_Preferred
The bi-directional transfer cap is used along with a 2L, 3L or 5L shake flask to connect a liquid inlet tubing with the required device.The liquid transfer is achieved by connection of a peristaltic pump between the shake flask and the device.Upon completion of the transfer, the transfer cap can be replaced with a vent cap for culture.Closed transfer reduces the risks of contamination during liquid transfer.The cap is molded and connected in an integrated manner, reducing the risks of leakage and media residues.A variety of tubing diameters are available and aseptic welding of liquid inlet tubing under normal conditions is supported.High-quality materials and smooth inner wall of the tubing provide an excellent transfer performance.Sterility level (SAL) is maintained at 10-6.Endotoxin-free, and no ingredients of animal origin.
Catalog No. 50-273-3453 Supplier Chemglass Life Sciences Supplier No. CGN2165685
Add to Cart
Add to Cart
The bi-directional transfer cap is used along with a 2L, 3L or 5L shake flask to connect a liquid inlet tubing with the required device.The liquid transfer is achieved by connection of a peristaltic pump between the shake flask and the device.Upon completion of the transfer, the transfer cap can be replaced with a vent cap for culture.Closed transfer reduces the risks of contamination during liquid transfer.The cap is molded and connected in an integrated manner, reducing the risks of leakage and media residues.A variety of tubing diameters are available and aseptic welding of liquid inlet tubing under normal conditions is supported.High-quality materials and smooth inner wall of the tubing provide an excellent transfer performance.Sterility level (SAL) is maintained at 10-6.Endotoxin-free, and no ingredients of animal origin.