Learn More
Alkali Scientific Bi-directional Liquid Transfer Cap System for 3L High Efficiency Erlenmeyer Flasks, White, TPE Tube (50cm 1/4" ID 3/8" OD), Vent Filter (0.22 μm Φ24mm), Sterile, 1/pk, 4/cs
SDP
Catalog No. 503027565
Encompass_Preferred
Our 3L High Efficiency Erlenmeyer Flask comes equipped with a Bi-directional Transfer Cap with a TPE Tube (50cm 1/4" ID 3/8" OD) and a Vent Filter (0.22 μm – 24mm). This sterile flask is ideal for various laboratory applications. The Bi-directional Transfer Cap facilitates easy and efficient transfers, while the TPE Tube ensures a secure connection. The Vent Filter, with a pore size of 0.22 μm, provides reliable filtration. Each pack contains 1 flask, and there are 4 packs per case. This setup is designed to streamline your workflow and enhance productivity in sterile environments. 
Catalog No. 50-302-7565 Supplier Alkali Scientific Supplier No. EFC621
Our 3L High Efficiency Erlenmeyer Flask comes equipped with a Bi-directional Transfer Cap with a TPE Tube (50cm 1/4" ID 3/8" OD) and a Vent Filter (0.22 μm – 24mm). This sterile flask is ideal for various laboratory applications. The Bi-directional Transfer Cap facilitates easy and efficient transfers, while the TPE Tube ensures a secure connection. The Vent Filter, with a pore size of 0.22 μm, provides reliable filtration. Each pack contains 1 flask, and there are 4 packs per case. This setup is designed to streamline your workflow and enhance productivity in sterile environments. 