Learn More
Selleck Chemical LLC BI6015-E0401-5MG
SDP
Supplier:  Selleck Chemical LLC E04015MG
Encompass_Preferred
BI6015 a hepatocyte nuclear factor 4 (HNF4 ) antagonist induces loss of HNF4 expression and hepatic steatosis in vivo BI6015 is cytotoxic to human hepatocellular carcinoma (HCC)
Catalog No. 50-238-9937