Learn More
Apexbio Technology LLC BI6727 (Volasertib)(Synonyms: Volasertib, BI-6727, BI 6727, Plk inhibitor BI6727, Polo-like kinase inhibitor BI6727), 10mM (in 1mL DMSO), CAS: 755038-65-4.
SDP
Supplier:  Apexbio Technology LLC A85585.1
Encompass_Preferred
BI6727 (Volasertib CAS 755038-65-4) is a selective inhibitor of Polo-like kinases (Plk) primarily targeting Plk1 with an IC50 of 0 87 nM and showing lower potency towards Plk2 (IC50 5 nM) and Plk3 (IC50 56 nM) Plk1 regulates the G2/M cell cycle transition and is commonly overexpressed in various tumor types representing a potential therapeutic target BI6727 inhibits proliferation across multiple tumor cell lines including colorectal (HCT116 EC50 23 nM) lung (NCI-H460 EC50 21 nM) melanoma (BRO EC50 11 nM) and hematologic cancer (GRANTA EC50 15 nM) In vivo BI6727 administration induced apoptosis and reduced tumor growth in mouse HCT116 and NCI-H460 xenograft models
Catalog No. 50-000-60110