Learn More
Medchemexpress LLC HY-120929 5mg Medchemexpress, BI8622 CAS:1875036-74-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1209295MG
Encompass_Preferred
Medchemexpress, HY-120929 5mg BI8622 CAS:1875036-74-0 BI8622 is a specific inhibitor of the ubiquitin ligase HUWE1 with an IC 50 of 3.1 μM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2553