Learn More
Selleck Chemical LLC BIA 10-2474
SDP
Catalog No. 5000002836
Encompass_Preferred
BIA 10-2474 is a novel fatty acid amide hydrolase (FAAH) inhibitor with a potent inhibitory effect upon FAAH and prolonged action in vivo
Catalog No. 50-000-02836 Supplier Selleck Chemical LLC Supplier No. S59325MG
BIA 10-2474 is a novel fatty acid amide hydrolase (FAAH) inhibitor with a potent inhibitory effect upon FAAH and prolonged action in vivo