Learn More
Medchemexpress LLC HY-19740 5mg Medchemexpress, BIA 10-2474 CAS:1233855-46-3 Purity:>98%
SDP
Catalog No. 502030877
Encompass_Preferred
Medchemexpress, HY-19740 5mg BIA 10-2474 CAS:1233855-46-3 BIA 10-2474 is an inhibitor of fatty acid amide hydrolase (FAAH) with IC50 values of 50 to 70mg/kg in various rat brain regions. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0877 Supplier Medchemexpress LLC Supplier No. HY197405MG
Medchemexpress, HY-19740 5mg BIA 10-2474 CAS:1233855-46-3 BIA 10-2474 is an inhibitor of fatty acid amide hydrolase (FAAH) with IC50 values of 50 to 70mg/kg in various rat brain regions. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.