Learn More
Medchemexpress LLC Biacetyl monoxime | 57-71-6 | MFCD00002116 | 98.0% | 101.10 g/mol | C4H7NO2 | 1g
SDP
Catalog No. 5000269717
Encompass_Preferred
Biacetyl monoxime (Diacetyl monoxime) a myosin ATPase inhibitor is a skeletal and cardiac muscle contraction inhibitor Biacetyl monoxime is also a well-characterized non-competitive inhibitor of chemical and motile activity of skeletal muscle myosin-II Biacetyl monoxime induces sarcoplasmic reticulum Ca2 release[1][2][3]
Catalog No. 50-002-69717 Supplier Medchemexpress LLC Supplier No. HYY04131G
Biacetyl monoxime (Diacetyl monoxime) a myosin ATPase inhibitor is a skeletal and cardiac muscle contraction inhibitor Biacetyl monoxime is also a well-characterized non-competitive inhibitor of chemical and motile activity of skeletal muscle myosin-II Biacetyl monoxime induces sarcoplasmic reticulum Ca2 release[1][2][3]