Learn More
Bioworld Bialaphos, 500 mg
Catalog No. 50255406
Encompass_Preferred
Naturally occurring antibiotic making it a particularly useful agent for selection in transformation research.Bialaphos is a natural herbicide produced by the bacteria Streptomyces hygroscopicus and Streptomyces viridochromegenes. Bialaphos is made of two alanine resudies and the glutamic acid analog phosphinothricin. It is commonly used as a gene selector in plants. Commonly used plasmids include: pGreenII 0229 and pGreenII 0229 62-SK.• Bialaphos Sodium Salt • Plant Tissue Culture TestedAppearance: SolidMelting Point: 160 °C
Catalog No. 50-255-406 Supplier Bioworld Supplier No. 402100012
Naturally occurring antibiotic making it a particularly useful agent for selection in transformation research.Bialaphos is a natural herbicide produced by the bacteria Streptomyces hygroscopicus and Streptomyces viridochromegenes. Bialaphos is made of two alanine resudies and the glutamic acid analog phosphinothricin. It is commonly used as a gene selector in plants. Commonly used plasmids include: pGreenII 0229 and pGreenII 0229 62-SK.• Bialaphos Sodium Salt • Plant Tissue Culture TestedAppearance: SolidMelting Point: 160 °C