Learn More
Gold Biotechnology Inc Bialaphos Sodium Salt 250 mg
SDP
Catalog No. 5000562174
Encompass_Preferred
Bialaphos is a tripeptide antibiotic that is used in transformation experiments of many species of plants that make use of the bar gene for selection In corn it is more effective than glufosinate In wheat bialaphos was the most reliable selection criteria used to identify bar transformed plants
Catalog No. 50-005-62174 Supplier Gold Biotechnology Inc Supplier No. B0178250
Bialaphos is a tripeptide antibiotic that is used in transformation experiments of many species of plants that make use of the bar gene for selection In corn it is more effective than glufosinate In wheat bialaphos was the most reliable selection criteria used to identify bar transformed plants