Learn More
Gold Biotechnology Inc Bialaphos Sodium Salt 500 mg
SDP
Catalog No. 5000562108
Encompass_Preferred
Bialaphos is a tripeptide antibiotic that is used in transformation experiments of many species of plants that make use of the bar gene for selection In corn it is more effective than glufosinate In wheat bialaphos was the most reliable selection criteria used to identify bar transformed plants
Catalog No. 50-005-62108 Supplier Gold Biotechnology Inc Supplier No. B0178500
Bialaphos is a tripeptide antibiotic that is used in transformation experiments of many species of plants that make use of the bar gene for selection In corn it is more effective than glufosinate In wheat bialaphos was the most reliable selection criteria used to identify bar transformed plants