Learn More
Bioworld Bialaphos, 1 mg/mL, Sterile, 25 mL
Catalog No. 50255408
Encompass_Preferred
Naturally occurring antibiotic making it a particularly useful agent for selection in transformation research.A tripeptide antibiotic employed in transformation experiments in plantsBialaphos Sodium Salt `58; Solution (1 mg/mL) `58; Plant Tissue Culture Tested `58; Sterile FilteredAppearance: LiquidMelting Point: 160 °C
Catalog No. 50-255-408 Supplier Bioworld Supplier No. 402100021
Naturally occurring antibiotic making it a particularly useful agent for selection in transformation research.A tripeptide antibiotic employed in transformation experiments in plantsBialaphos Sodium Salt `58; Solution (1 mg/mL) `58; Plant Tissue Culture Tested `58; Sterile FilteredAppearance: LiquidMelting Point: 160 °C