Learn More
Cayman Chemical Bialaphos (sodium salt), Purity: ≥95%, CAS: 71048-99-2, 50 mg
SDP
Catalog No. 502965219
Encompass_Preferred
A natural non-selective phytotoxin that is converted in vivo to the active agent phosphinothricin, which inhibits glutamine synthetase (Ki = 6.1 µM); used in the selection of transgenic plants that express the bar gene, usually under the control of a constitutively active viral promoter
Catalog No. 50-296-5219 Supplier Cayman Chemical Supplier No. 1675450 MG
A natural non-selective phytotoxin that is converted in vivo to the active agent phosphinothricin, which inhibits glutamine synthetase (Ki = 6.1 µM); used in the selection of transgenic plants that express the bar gene, usually under the control of a constitutively active viral promoter