Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biapenem | 120410-24-4 | MFCD00864863 | 98.0% | C15H18N4O4S | 10MG
SDP

Catalog No. 501872914
Encompass_Preferred

Biapenem is a carbapenem-class antibacterial agent provided as an analytical standard for research and analytical applications. It exhibits broad-spectrum activity and is supplied with specified purity and characterization for use in assays, method development, and reference standards.

  • Carbapenem-class antibacterial agent with broad-spectrum activity.
  • Intended for research and analytical applications.
  • Purity 98.0%.
  • Chemical formula C15H18N4O4S and molecular weight 350.39 g/mol.
  • Provided as a small-format analytical standard suitable for assay and reference use.
  • CAS number 120410-24-4 for unambiguous identification.

Catalog No. 50-187-2914 Supplier Medchemexpress LLC Supplier No. HY1357310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biapenem is a carbapenem-class antibacterial agent provided as an analytical standard for research and analytical applications. It exhibits broad-spectrum activity and is supplied with specified purity and characterization for use in assays, method development, and reference standards.

  • Carbapenem-class antibacterial agent with broad-spectrum activity.
  • Intended for research and analytical applications.
  • Purity 98.0%.
  • Chemical formula C15H18N4O4S and molecular weight 350.39 g/mol.
  • Provided as a small-format analytical standard suitable for assay and reference use.
  • CAS number 120410-24-4 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.