Learn More
Apexbio Technology LLC Biapenem 120410-24-4 100mg
SDP
Catalog No. 5000096323
Encompass_Preferred
Biapenem (120410-24-4) is a small-molecule inhibitor targeting penicillin-binding proteins (PBPs) It is designed to inhibit bacterial cell wall synthesis thereby disrupting cell wall integrity and leading to bacterial cell death Biapenem exerts its biological activity primarily through binding to and inhibiting PBPs In in vitro studies biapenem demonstrates inhibitory activity with IC50 values ranging from 0 1 to 10 g/mL depending on organism susceptibility and assay conditions Based on these pharmacological properties biapenem holds research potential in susceptibility testing antimicrobial combination studies and exploration of bacterial resistance mechanisms
Catalog No. 50-000-96323 Supplier Apexbio Technology LLC Supplier No. A1373100
Biapenem (120410-24-4) is a small-molecule inhibitor targeting penicillin-binding proteins (PBPs) It is designed to inhibit bacterial cell wall synthesis thereby disrupting cell wall integrity and leading to bacterial cell death Biapenem exerts its biological activity primarily through binding to and inhibiting PBPs In in vitro studies biapenem demonstrates inhibitory activity with IC50 values ranging from 0 1 to 10 g/mL depending on organism susceptibility and assay conditions Based on these pharmacological properties biapenem holds research potential in susceptibility testing antimicrobial combination studies and exploration of bacterial resistance mechanisms