Learn More
Grainger CONDOR Bib Apron, PVC
Catalog No. NC1064559
Encompass_Preferred
The Condor Bib Apron PVC is a protective apron made of 0.5mm clear PVC. It features a bib-style design with adjustable neck and waist straps for a customized fit. The apron provides protection against dirt, grease, and liquid splashes on the front of the body from the chest to the knees. It can be wiped clean for repeated use. This durable and reusable apron is ideal for use in industrial, food service, and laboratory settings.
This item is not returnable. View return policy
Catalog No. NC1064559 Supplier Grainger Supplier No. 3XE79
The Condor Bib Apron PVC is a protective apron made of 0.5mm clear PVC. It features a bib-style design with adjustable neck and waist straps for a customized fit. The apron provides protection against dirt, grease, and liquid splashes on the front of the body from the chest to the knees. It can be wiped clean for repeated use. This durable and reusable apron is ideal for use in industrial, food service, and laboratory settings.