Learn More
Grainger North Primair 100 Bib Hood, Universal Size, 3/Bx.
Catalog No. 502794420
Encompass_Preferred
These replacement bib hoods are designed for use with the Primair 100 series respiratory protection systems. They are constructed from spun bond polypropylene with a polyethylene coating and feature a clear co-polyester visor for optimal visibility. The universal sizing accommodates a wide range of face shapes and sizes, ensuring reliable protection in various industrial applications.
  • NIOSH approved when used with compatible air systems
  • Includes 1 set of 3 mylar peel-away cover lens
  • Latex free construction
  • Adjustable headgear and length for customizable comfort
  • Assigned protection factor (APF) of 1,000
  • Packaging: 3 per box
Catalog No. 50-279-4420 Supplier Grainger Supplier No. 1AAP4
These replacement bib hoods are designed for use with the Primair 100 series respiratory protection systems. They are constructed from spun bond polypropylene with a polyethylene coating and feature a clear co-polyester visor for optimal visibility. The universal sizing accommodates a wide range of face shapes and sizes, ensuring reliable protection in various industrial applications.
  • NIOSH approved when used with compatible air systems
  • Includes 1 set of 3 mylar peel-away cover lens
  • Latex free construction
  • Adjustable headgear and length for customizable comfort
  • Assigned protection factor (APF) of 1,000
  • Packaging: 3 per box